(±)-Tetrahydrofurfurylbromid

Lieferant: Thermo Scientific

Synonyme: 2-(Bromomethyl)tetrahydrofuran

CC16908.10 CC16908.50
MAYBCC16908.10EA 0 EUR
MAYBCC16908.10 MAYBCC16908.50
(±)-Tetrahydrofurfurylbromid
(±)-Tetrahydrofurfurylbromid
Formel: C₅H₉BrO
Molecular Weight: 165,04 g/mol
MDL: MFCD00016894
CAS-Nummer: 1192-30-9
EINECS: 214-750-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR