Ammoniumchlorid 250 g/l analytisches Reagens
Lieferant: Bernd Kraft
Synonyme:
Salmiak
10700.2000
10700.3000
10700.5020
KRAF10700.2000EA
0
EUR
KRAF10700.2000
KRAF10700.3000
KRAF10700.5020
Ammoniumchlorid 250 g/l analytisches Reagens
Ammoniumchlorid
Formel:
NH₄Cl Dichte: 1,073 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011420 CAS-Nummer: 12125-02-9 EINECS: 235-186-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...