6-Azauracil ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1,2,4-Triazine-3,5(2H,4H)-dione
A14389.14
A14389.06
A14389.14EA
178
EUR
A14389.14
A14389.06
6-Azauracil ≥98%
6-Azauracil
Purity: 98%
CAS No.: 461-89-2
Molecular Formula: C3H3N3O2
Formula Weight: 113.08
Melting Point: 274-282°
Beilstein Reference: 116472
Formel:
C₃H₃N₃O₂ Schmelzpunkt: 274…282 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006456 CAS-Nummer: 461-89-2 EINECS: 207-318-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...