N,N-Diethyl-2-ethanolammoniumchlorid zur Synthese, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N-Diethylethanolammonium chloride

M-Clarity™ Qualitätsstufe = MQ200

8.20394.0250 8.20394.1000
8.20394.0250EA 0 EUR
8.20394.0250 8.20394.1000
N,N-Diethyl-2-ethanolammoniumchlorid zur Synthese, Sigma-Aldrich®
N,N-Diethyl-2-ethanolammoniumchlorid
Formel: C₆H₁₄ClNO
CAS-Nummer: 13989-32-7
EINECS: 237-787-7

Bestellen

Spezifikation Testergebnisse

Solubility (20 °C) soluble
Melting Point 133 - 136 °C
Molar Mass 153.65 g/mol
Synonyms N,N-Diethyl-2-hydroxyethylammonium chloride, 2-Diethylaminoethanol hydrochloride
Hill Formula C6H16ClNO
Chemical Formula (C2H5)2NCH2CH2OH * HCl


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR