(±)-Tetrahydrofuran-2-carbonsäure 98+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Tetrahydrofuran-2-carboxylic acid, 2-Tetrahydrofuran Carboxylic Acid
263141000
263145000
263140250
ACRO263141000EA
97.4
EUR
ACRO263141000
ACRO263145000
ACRO263140250
(±)-Tetrahydrofuran-2-carbonsäure 98+%
(±)-Tetrahydrofuran-2-carbonsäure
Formel:
C₅H₈O₃ Molecular Weight: 116.12 g/mol Siedepunkt: 128…129 °C (13 mmHg) Dichte: 1.17 g/cm³ Flammpunkt: 139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022473 CAS-Nummer: 16874-33-2 UN: 3265 ADR: 8 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4580 to 1.4620 (20°C, 589 nm) |
GC | ≥98.0 % |
Titration with NaOH | 98.0 to 102.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...