Rhodamin 6G Perchlorat, Sigma-Aldrich®

Lieferant: Merck

252441-250MG 252441-1G
SIAL252441-250MGEA 45.9 EUR
SIAL252441-250MG SIAL252441-1G
Rhodamin 6G Perchlorat, Sigma-Aldrich®
Rhodamin 6G Perchlorat
Formel: C₂₈H₃₁ClN₂O₇
Molecular Weight: 543,02 g/mol
MDL: MFCD00013117
CAS-Nummer: 13161-28-9
EINECS: 236-099-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR