(2-Nitrophenyl)-β-D-galactopyranosid (ONPG), Sigma-Aldrich®
Lieferant: Merck
Synonyme: o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
M-Clarity™ Qualitätsstufe = MQ200
N1127-1G N1127-250MG N1127-25G N1127-500MG N1127-5G
SIALN1127-1GEA 56.4 EUR
SIALN1127-1G SIALN1127-250MG SIALN1127-25G SIALN1127-500MG SIALN1127-5G
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG), Sigma-Aldrich®
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG)
Formel: C₁₂H₁₅NO₈
Lagertemperatur: Gefrierschrank
MDL: MFCD00063255
CAS-Nummer: 369-07-3
EINECS: 206-716-1