(4-Nitrophenyl)-α-D-galactopyranosid ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside

L13376.MC L13376.ME
L13376.MCEA 24.5 EUR
L13376.MC L13376.ME
(4-Nitrophenyl)-α-D-galactopyranosid ≥98%
(4-Nitrophenyl)-α-D-galactopyranosid

Purity: 98+% CAS No.: 7493-95-0 Molecular Formula: C12H15NO8 Formula Weight: 301.26 Storage Temperature: 2°C to 8°C Melting Point: 166-169° Beilstein Reference: 92214 Optical Rotation: +242° (c=1 in water) MDL No.: MFCD00065050

Formel: C₁₂H₁₅NO₈
Schmelzpunkt: 166…169 °C
MDL: MFCD00065050
CAS-Nummer: 7493-95-0
EINECS: 231-343-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR