(4-Nitrophenyl)-α-D-galactopyranosid, Pulver
Lieferant: MP Biomedicals
Synonyme:
(2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside
0210247105
02102471.2
02102471.5
0210247101
ICNA0210247105EA
812
EUR
ICNA0210247105
ICNA02102471.2
ICNA02102471.5
ICNA0210247101
(4-Nitrophenyl)-α-D-galactopyranosid, Pulver
(4-Nitrophenyl)-α-D-galactopyranosid
p-Nitrophenyl-alpha-D-galactopyranoside is used as a substrate for α-galactosidase.
Soluble in water (5% w/v), HCl (50 mg/ml - clear, colourless solution) & methanol (100 mg/ml).
Formel:
C₁₂H₁₅NO₈ Molecular Weight: 301.26 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00065050 CAS-Nummer: 7493-95-0 EINECS: 231-343-6 |
Spezifikation Testergebnisse
Identity Test | Passes |
FTIR | Conforms to Standard C |
Melting Point | 164 - 176°C |
Optical Rotation (c=1, 1N HCl) | +240° ± 20° |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...