(4-Nitrophenyl)-α-D-galactopyranosid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2R,3R,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol, 4-Nitrophenyl α-D-galactopyranoside

M-Clarity™ Qualitätsstufe = MQ200

N0877-100MG N0877-1G N0877-250MG N0877-500MG N0877-5G
SIALN0877-100MGEA 57.4 EUR
SIALN0877-100MG SIALN0877-1G SIALN0877-250MG SIALN0877-500MG SIALN0877-5G
(4-Nitrophenyl)-α-D-galactopyranosid, Sigma-Aldrich®
(4-Nitrophenyl)-α-D-galactopyranosid
Formel: C₁₂H₁₅NO₈
MDL: MFCD00065050
CAS-Nummer: 7493-95-0
EINECS: 231-343-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR