(4-Nitrophenyl)-β-D-galactopyranosid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: pNPG, 4-Nitrophenyl β-D-Galactopyranoside

M-Clarity™ Qualitätsstufe = MQ300

N1252-10G N1252-1G N1252-250MG N1252-500MG N1252-5G
SIALN1252-10GEA 1750 EUR
SIALN1252-10G SIALN1252-1G SIALN1252-250MG SIALN1252-500MG SIALN1252-5G
(4-Nitrophenyl)-β-D-galactopyranosid, Sigma-Aldrich®
(4-Nitrophenyl)-β-D-galactopyranosid
Formel: C₁₂H₁₅NO₈
Molecular Weight: 301,25 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00063256
CAS-Nummer: 3150-24-1
EINECS: 221-584-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR