(3-Indoxyl)-β-D-glucopyranosid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indoxyl-D-glucoside, 3-Indole-D-glucoside, 3-Indolyl-D-glucoside, Indoxyl-B-D Glucoside, Indican (Plant Indican)

M-Clarity™ Qualitätsstufe = MQ200

I3750-100MG I3750-1G I3750-25MG I3750-2G I3750-500MG
SIALI3750-100MGEA 207 EUR
SIALI3750-100MG SIALI3750-1G SIALI3750-25MG SIALI3750-2G SIALI3750-500MG
(3-Indoxyl)-β-D-glucopyranosid, Sigma-Aldrich®
(3-Indoxyl)-β-D-glucopyranosid
Formel: C₁₄H₁₇NO₆
Lagertemperatur: Gefrierschrank
MDL: MFCD00047169
CAS-Nummer: 487-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR