1-Naphthylphosphat Mononatriumsalz Monohydrat
Lieferant: Serva
|
Achtung
|
Synonyme:
1-Naphthylphosphoric Acid Monosodium Salt Monohydrate, Monosodium 1-Naphthyl Phosphate Monohydrate, Sodium 1-Naphthyl Hydrogen Phosphate Monohydrate, Phosphoric Acid 1-Naphthyl Ester Monosodium Salt Monohydrate
30130.02
30130.03
SERA30130.02EA
0
EUR
SERA30130.02
SERA30130.03
1-Naphthylphosphat Mononatriumsalz Monohydrat
1-Naphthylphosphat Mononatriumsalz Monohydrat
Formel:
C₁₀H₈NaO₄P·1H₂O Molecular Weight: 264,15 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00150615 CAS-Nummer: 81012-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...