1-Naphthylphosphat Dinatriumsalz Monohydrat ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Naphthyl phosphate, disodium salt hydrate
B22866.03
B22866.06
B22866.14
B22866.03EA
57
EUR
B22866.03
B22866.06
B22866.14
1-Naphthylphosphat Dinatriumsalz Monohydrat ≥99%
1-Naphthylphosphat Dinatriumsalz Monohydrat
Purity: 99%
CAS No.: 207569-06-0
Molecular Formula: C10H7Na2O4P.xH2O
Formula Weight: 268.12(anhy)
Storage Temperature: 2°C to 8°C
Melting Point:
Formel:
C₁₀H₇Na₂O₄P·1H₂O Molecular Weight: 268,12 g/mol (anhy) Schmelzpunkt: >300 °C |
MDL:
MFCD00150614 CAS-Nummer: 207569-06-0 EINECS: 218-564-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...