(±)-2-Methylglutarsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methylpentanedioic acid

A11823.06 A11823.14 A11823.22
A11823.06EA 54.7 EUR
A11823.06 A11823.14 A11823.22
(±)-2-Methylglutarsäure ≥98%
(±)-2-Methylglutarsäure

Purity: 98% CAS No.: 617-62-9 Molecular Formula: C6H10O4 Formula Weight: 146.14 Melting Point: 74-78° Boiling Point: 214-215°/22mm MDL No.: MFCD00002661

Formel: C₆H₁₀O₄
Molecular Weight: 146,14 g/mol
Siedepunkt: 214…215 °C (22 mmHg)
Schmelzpunkt: 74…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002661
CAS-Nummer: 617-62-9
EINECS: 210-521-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR