N-Decanoyl-N-methyl-D-glucamin (MEGA-10)

Lieferant: BACHEM BIOCHEMICA

4011133.0001 4011133.0005 4011133.0025
BACE4011133.0001EA 0 EUR
BACE4011133.0001 BACE4011133.0005 BACE4011133.0025
N-Decanoyl-N-methyl-D-glucamin (MEGA-10)
N-Decanoyl-N-methyl-D-glucamin (MEGA-10)
Formel: C₁₇H₃₅NO₆
Molecular Weight: 349.46 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00036801
CAS-Nummer: 85261-20-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR