N-Methyl-N-octanoyl-D-glucamin (MEGA-8)

Lieferant: BACHEM BIOCHEMICA

Synonyme: N-Octanoyl-N-methyl-D-glucamine, N-Methyl-N-((2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl)octanamide, MEGA-8

4011132.0001 4011132.0005
BACE4011132.0001EA 0 EUR
BACE4011132.0001 BACE4011132.0005
N-Methyl-N-octanoyl-D-glucamin (MEGA-8)
N-Methyl-N-octanoyl-D-glucamin (MEGA-8)
Formel: C₁₅H₃₁NO₆
Molecular Weight: 321.4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134152
CAS-Nummer: 85316-98-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR