L(+)-Glutaminsäure Hydrochlorid ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
(S)-2-Aminopentanedioic acid hydrochloride, 2-Aminoglutaric acid hydrochloride
A12505.0B
A12505.30
A12505.0BEA
108
EUR
A12505.0B
A12505.30
L(+)-Glutaminsäure Hydrochlorid ≥99%
L(+)-Glutaminsäure Hydrochlorid
Purity: 99%
CAS No.: 138-15-8
Molecular Formula: C5H9NO4.HCl
Formula Weight: 183.59
Melting Point: 203° dec.
Beilstein Reference: 3565569
Merck Reference: 14,4469
Optical Rotation: +23° (c=6 in water)
MDL No.: MFCD00012619
Formel:
C₅H₁₀ClNO₄ Molecular Weight: 183,59 g/mol Schmelzpunkt: 203 °C (Zers.) Dichte: 1,525 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012619 CAS-Nummer: 138-15-8 EINECS: 205-315-9 Merck Index: 14,4469 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...