Creatinphosphat Dinatriumsalz Tetrahydrat ≥98%, weißes Pulver

Lieferant: MP Biomedicals

Synonyme: Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate

0210052025 0210052001 0210052005
ICNA0210052025EA 972 EUR
ICNA0210052025 ICNA0210052001 ICNA0210052005
Creatinphosphat Dinatriumsalz Tetrahydrat ≥98%, weißes Pulver
Creatinphosphat Dinatriumsalz Tetrahydrat

Creatine phosphate is substrate for CPK determination.


  • Substantially free of creatine


Substantially free of creatine.


Soluble in water (150 mg/ml-clear, colorless solution)

Formel: C₄H₈N₃Na₂O₅P·4H₂O
Molecular Weight: 327,14 g/mol (255,07 anhy)
Lagertemperatur: Gefrierschrank
MDL: MFCD00150192
CAS-Nummer: 71519-72-7
EINECS: 213-074-6

Bestellen

Spezifikation Testergebnisse

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
pH (1% aq soln) 7 - 9
Moisture 22 ± 3%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR