Creatinphosphat Dinatriumsalz Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Phosphocreatine disodium hydrate, Sodium 2-(1-methyl-3-phosphonatoguanidino)acetate hydrate(2:1:x), N-(Imino[phosphonoamino]methyl)-N-methylglycine hydrate, Phosphocreatine disodium salt hydrate
M-Clarity™ Qualitätsstufe = MQ200
P7936-10MG
P7936-1G
P7936-25G
P7936-5G
SIALP7936-10MGEA
41.2
EUR
SIALP7936-10MG
SIALP7936-1G
SIALP7936-25G
SIALP7936-5G
Creatinphosphat Dinatriumsalz Hydrat, Sigma-Aldrich®
Creatinphosphat Dinatriumsalz Hydrat
Formel:
C₄H₈N₃Na₂O₅P·H₂O Molecular Weight: 273.09 g/mol |
MDL:
MFCD00150191 CAS-Nummer: 19333-65-4 EINECS: 213-074-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...