Tris-acetat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate

93337-100G 93337-25G
SIAL93337-100GEA 298 EUR
SIAL93337-100G SIAL93337-25G
Tris-acetat, Sigma-Aldrich®
Tris-acetat
Formel: NH₂C(CH₂OH)₃·CH₃COOH
Lagertemperatur: Raumtemperatur
MDL: MFCD00038951
CAS-Nummer: 6850-28-8
EINECS: 229-939-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR