Tris-acetat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
M-Clarity™ Qualitätsstufe = MQ200
T1258-100G
T1258-250G
T1258-25G
SIALT1258-100GEA
275
EUR
SIALT1258-100G
SIALT1258-250G
SIALT1258-25G
Tris-acetat, Sigma-Aldrich®
Tris-acetat
Formel:
NH₂C(CH₂OH)₃·CH₃COOH Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038951 CAS-Nummer: 6850-28-8 EINECS: 229-939-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...