N-Methylbistrifluoracetamid

Lieferant: FLUOROCHEM
Gefahr

Synonyme: 2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA

009279-25G 009279-100G
FLUO009279-25GEA 0 EUR
FLUO009279-25G FLUO009279-100G
N-Methylbistrifluoracetamid
N-Methylbistrifluoracetamid
Formel: C₅H₃F₆NO₂
Molecular Weight: 223,07 g/mol
MDL: MFCD00000412
CAS-Nummer: 685-27-8
EINECS: 211-680-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR