N-Methylbistrifluoracetamid Acylierungsreagenz, zur GC-Derivatisierung
Lieferant: MACHEREY-NAGEL
Gefahr
Synonyme: 2,2,2-Trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide, MBTFA
701410.110 701410.201 701410.510
MANA701410.110EA 170 EUR
MANA701410.110 MANA701410.201 MANA701410.510
N-Methylbistrifluoracetamid Acylierungsreagenz, zur GC-Derivatisierung
N-Methylbistrifluoracetamid
This derivitisation reagent is recommended for alcohols, primary and secondary amines as well as for thiols under mild, neutral conditions. MBTFA also forms very volatile derivatives with carbohydrates.
  • Addition of halogenated carbons enhances detectability by GC-ECD
  • Derivatives are hydrolytical stable
  • Increased sensitivity by adding molecular weight
Formel: C₅H₃F₆NO₂
Molecular Weight: 223,07 g/mol
Siedepunkt: 123…124 °C (760 mm Hg)
Dichte: 1,55 g/cm³ (d20°/4°)"
MDL: MFCD00000412
CAS-Nummer: 685-27-8
EINECS: 211-680-5
UN: 3316
ADR: 9,II