Bistrifluoracetamid

Lieferant: FLUOROCHEM

008535-5G 008535-25G
FLUO008535-5GEA 272 EUR
FLUO008535-5G FLUO008535-25G
Bistrifluoracetamid
Bistrifluoracetamid
Formel: C₄HF₆NO₂
MDL: MFCD00013564
CAS-Nummer: 407-24-9
EINECS: 206-981-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR