N,N-Diethyl-2,2,2-trifluoracetamid

Lieferant: FLUOROCHEM

013905-5G 013905-25G
FLUO013905-5GEA 121 EUR
FLUO013905-5G FLUO013905-25G
N,N-Diethyl-2,2,2-trifluoracetamid
N,N-Diethyl-2,2,2-trifluoracetamid
Formel: C₆H₁₀F₃NO
Molecular Weight: 169,15 g/mol
MDL: MFCD00192034
CAS-Nummer: 360-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR