N,N-Dimethyl-2,2,2-trifluoracetamid

Lieferant: FLUOROCHEM

Synonyme: 2,2,2-Trifluoro-N,N-dimethylacetamide

010299-5G 010299-1G 010299-100G 010299-25G
FLUO010299-5GEA 45.9 EUR
FLUO010299-5G FLUO010299-1G FLUO010299-100G FLUO010299-25G
N,N-Dimethyl-2,2,2-trifluoracetamid
N,N-Dimethyl-2,2,2-trifluoracetamid
Formel: C₄H₆F₃NO
Molecular Weight: 141,0900 g/mol
MDL: MFCD00043555
CAS-Nummer: 1547-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR