N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)

Lieferant: FLUOROCHEM

Synonyme: N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide, MBDSTFA

009280-5G 009280-25G 009280-100G
FLUO009280-5GEA 0 EUR
FLUO009280-5G FLUO009280-25G FLUO009280-100G
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
Formel: C₉H₁₈F₃NOSi
Molecular Weight: 241.33 g/mol
MDL: MFCD00009671
CAS-Nummer: 77377-52-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR