N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide, MBDSTFA

M-Clarity™ Qualitätsstufe = MQ200

394882-100ML 394882-10X1ML 394882-25ML 394882-5ML
SIAL394882-100MLEA 1110 EUR
SIAL394882-100ML SIAL394882-10X1ML SIAL394882-25ML SIAL394882-5ML
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA), Sigma-Aldrich®
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
Formel: C₉H₁₈F₃NOSi
Molecular Weight: 241.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009671
CAS-Nummer: 77377-52-7
UN: 1993
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR