N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
Lieferant: CHEMPUR SILANE
Synonyme:
N-(tert-Butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide, MBDSTFA
SILS11050.5
SILS11050.25
SILS11050.100
SILNSILS11050.5EA
0
EUR
SILNSILS11050.5
SILNSILS11050.25
SILNSILS11050.100
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
Formel:
C₉H₁₈F₃NOSi Molecular Weight: 241.33 g/mol |
MDL:
MFCD00009671 CAS-Nummer: 77377-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...