2-Mercaptoethansulfonsäure Natriumsalz (Mesna), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium 2-mercaptoethanesulphonate

63705-10G 63705-50G
SIAL63705-10GEA 161 EUR
SIAL63705-10G SIAL63705-50G
2-Mercaptoethansulfonsäure Natriumsalz (Mesna), Sigma-Aldrich®
2-Mercaptoethansulfonsäure Natriumsalz (Mesna)
Formel: C₂H₅NaO₃S₂
Molecular Weight: 164,18 g/mol
MDL: MFCD00007535
CAS-Nummer: 19767-45-4
EINECS: 243-285-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR