N-Cbz-Glycin (Z-Gly-OH) 98.5%

Lieferant: Thermo Scientific

Synonyme: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

108350250 108351000 108355000
ACRO108350250EA 35.6 EUR
ACRO108350250 ACRO108351000 ACRO108355000
N-Cbz-Glycin (Z-Gly-OH) 98.5%
N-Cbz-Glycin (Z-Gly-OH)
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,2 g/mol
Schmelzpunkt: 117…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002691
CAS-Nummer: 1138-80-3
EINECS: 214-516-0

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 122 °C
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %
Solubility light yellow solution
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR