N-Cbz-Glycin (Z-Gly-OH) ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid

A15928.14 A15928.22
A15928.14EA 37.5 EUR
A15928.14 A15928.22
N-Cbz-Glycin (Z-Gly-OH) ≥98%
N-Cbz-Glycin (Z-Gly-OH)

Purity: 98+% CAS No.: 1138-80-3 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Storage Temperature: 2°C to 8°C Melting Point: 119-122° Beilstein Reference: 526877 MDL No.: MFCD00002691

Formel: C₁₀H₁₁NO₄
Molecular Weight: 209.2 g/mol
Schmelzpunkt: 119…122 °C
MDL: MFCD00002691
CAS-Nummer: 1138-80-3
EINECS: 214-516-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR