(3-Methoxyphenyl)acetylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(3-Methoxyphenyl)acetyl chloride

M-Clarity™ Qualitätsstufe = MQ100

316938-25G 316938-5G
SIAL316938-25GEA 425 EUR
SIAL316938-25G SIAL316938-5G
(3-Methoxyphenyl)acetylchlorid, Sigma-Aldrich®
(3-Methoxyphenyl)acetylchlorid
Formel: C₉H₉ClO₂
Molecular Weight: 184.6196 g/mol
MDL: MFCD00012244
CAS-Nummer: 6834-42-0
UN: 3265
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR