3-Brombenzanilid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-N-phenylbenzamide

CA-4485-5G CA-4485-1G
COBBCA-4485-5GEA 399 EUR
COBBCA-4485-5G COBBCA-4485-1G
3-Brombenzanilid
3-Brombenzanilid
Formel: C₁₃H₁₀BrNO
Molecular Weight: 276,1300 g/mol
MDL: MFCD00087947
CAS-Nummer: 63710-33-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR