2-Brom-6-fluor-4-nitrophenol
Lieferant: COMBI-BLOCKS
AN-1554-5G
AN-1554-1G
COBBAN-1554-5GEA
204
EUR
COBBAN-1554-5G
COBBAN-1554-1G
2-Brom-6-fluor-4-nitrophenol
2-Brom-6-fluor-4-nitrophenol
Formel:
C₆H₃BrFNO₃ Molecular Weight: 236,00 g/mol |
MDL:
MFCD09907952 CAS-Nummer: 329-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...