2-Brom-4-fluor-6-nitrophenol

Lieferant: COMBI-BLOCKS

AN-1331-5G AN-1331-25G AN-1331-1G
COBBAN-1331-5GEA 0 EUR
COBBAN-1331-5G COBBAN-1331-25G COBBAN-1331-1G
2-Brom-4-fluor-6-nitrophenol
2-Brom-4-fluor-6-nitrophenol
Formel: C₆H₃BrFNO₃
MDL: MFCD03094182
CAS-Nummer: 320-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR