1-Methylimidazoliumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Methyl-1H-imidazol-3-ium chloride

40477-1KG-F 40477-100G-F
SIAL40477-1KG-FEA 960 EUR
SIAL40477-1KG-F SIAL40477-100G-F
1-Methylimidazoliumchlorid, Sigma-Aldrich®
1-Methylimidazolium chloride
Formel: C₄H₇ClN₂
Molecular Weight: 118,5648 g/mol
MDL: MFCD06798176
CAS-Nummer: 35487-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR