2-(4-Morpholinyl)phenol
Lieferant: Thermo Scientific
Synonyme:
2-Morpholinophenol
S08696DA
S08696EA
MAYBS08696DAEA
0
EUR
MAYBS08696DA
MAYBS08696EA
2-(4-Morpholinyl)phenol
2-(4-Morpholinyl)phenol
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179,215 g/mol |
MDL:
MFCD00051674 CAS-Nummer: 41536-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...