3-(4-Morpholinyl)phenol

Lieferant: Thermo Scientific

Synonyme: 3-Morpholinophenol

TL00276DA TL00276EA
MAYBTL00276DAEA 0 EUR
MAYBTL00276DA MAYBTL00276EA
3-(4-Morpholinyl)phenol
3-(4-Morpholinyl)phenol
Formel: C₁₀H₁₃NO₂
Molecular Weight: 179.22 g/mol
MDL: MFCD00051675
CAS-Nummer: 27292-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR