3,3'-Diaminobenzidin Tetrahydrochlorid (DAB)
Lieferant: Serva
Synonyme: 3,3',4,4'-Biphenyltetramine tetrahydrochloride, DAB HCl
18865.01 18865.02
SERA18865.01EA 0 EUR
SERA18865.01 SERA18865.02
3,3'-Diaminobenzidin Tetrahydrochlorid (DAB)
3,3'-Diaminobenzidin Tetrahydrochlorid
Formel: C₁₂H₁₈Cl₄N₄
Molecular Weight: 360,11 g/mol
MDL: MFCD00012969
CAS-Nummer: 7411-49-6
EINECS: 231-018-9