4-Nitrochinolin-N-oxid

Lieferant: CHEMPUR

JFD03905.250 JFD03905.1
CHMPJFD03905.250EA 0 EUR
CHMPJFD03905.250 MAYBJFD03905.1
4-Nitrochinolin-N-oxid
4-Nitrochinolin-N-oxid
Formel: C₉H₆N₂O₃
MDL: MFCD00006738
CAS-Nummer: 56-57-5
EINECS: 200-281-1
UN: 2811
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR