Zinn(II)methanolat

Lieferant: Thermo Scientific
Gefahr

20897.06 20897.14
20897.06EA 114 EUR
20897.06 20897.14
Zinn(II)methanolat
Zinn(II)methanolat

CAS No.: 14794-99-1 Molecular Formula: Sn(OCH3)2 Formula Weight: 180.76 Physical Form: Powder Sensitivity: Moisture Sensitive Melting Point: 242-243° MDL No.: MFCD00015612

Formel: C₂H₆O₂Sn
Molecular Weight: 180,76 g/mol
Schmelzpunkt: 242…243 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015612
CAS-Nummer: 14794-99-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR