Naphthol AS-TR-phosphat Dinatriumsalz, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ300
N6125-100MG
N6125-1G
N6125-5G
SIALN6125-100MGEA
55.1
EUR
SIALN6125-100MG
SIALN6125-1G
SIALN6125-5G
Naphthol AS-TR-phosphat Dinatriumsalz, Sigma-Aldrich®
Naphthol AS-TR-phosphat Dinatriumsalz
Formel:
C₁₈H₁₃ClNNa₂O₅P |
CAS-Nummer:
4264-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...