1-Naphthylphosphat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

N5602-1G N5602-5G
SIALN5602-1GEA 62.7 EUR
SIALN5602-1G SIALN5602-5G
1-Naphthylphosphat, Sigma-Aldrich®
1-Naphthylphosphat
Formel: C₁₀H₉O₄P
CAS-Nummer: 1136-89-6
EINECS: 214-502-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR