1-Naphthylphosphat Dinatriumsalz Monohydrat, kristallin

Lieferant: MP Biomedicals
Achtung

Synonyme: 1-Naphthyl phosphate, disodium salt hydrate

02100257.1 02100257.2 0210025701 0210025705
ICNA02100257.1EA 33.9 EUR
ICNA02100257.1 ICNA02100257.2 ICNA0210025701 ICNA0210025705
1-Naphthylphosphat Dinatriumsalz Monohydrat, kristallin
1-Naphthylphosphat Dinatriumsalz Monohydrat

α - Naphthyl phosphate disodium salt is a substrate for determination of phosphatase.


1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.


Soluble in water (50 mg/ml)

Formel: C₁₀H₇Na₂O₄P·1H₂O
Molecular Weight: 268,12 g/mol (anhy)
Schmelzpunkt: 157…159 °C (Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00150614
CAS-Nummer: 207569-06-0
EINECS: 218-564-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR