1-Naphthylphosphat Dinatriumsalz, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Naphthylphosphoric acid disodium salt, α-Naphthyl Phosphate Disodium Salt Hydrate, Phosphoric acid 1-Naphthyl ester disodium salt, Disodium 1-Naphthyl phosphate hydrate

M-Clarity™ Qualitätsstufe = MQ200

N7255-100MG N7255-1G N7255-250MG N7255-5G
SIALN7255-100MGEA 43.7 EUR
SIALN7255-100MG SIALN7255-1G SIALN7255-250MG SIALN7255-5G
1-Naphthylphosphat Dinatriumsalz, Sigma-Aldrich®
1-Naphthylphosphat Dinatriumsalz
Formel: C₁₀H₇Na₂O₄P
MDL: MFCD00041007
CAS-Nummer: 2183-17-7
EINECS: 218-564-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR