Guanosin-5'-diphosphat Dinatriumsalz, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
((2R,3S,4R,5R)-5-(2-Amino-6-oxo-3,6-dihydro-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate, disodium salt, Guanosine 5'-diphosphate disodium salt
51060-100MG
51060-500MG
SIAL51060-100MGEA
151
EUR
SIAL51060-100MG
SIAL51060-500MG
Guanosin-5'-diphosphat Dinatriumsalz, Sigma-Aldrich®
Guanosin-5'-diphosphat Dinatriumsalz
Formel:
C₁₀H₁₃N₅Na₂O₁₁P₂ Molecular Weight: 487,17 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00070121 CAS-Nummer: 7415-69-2 EINECS: 231-026-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...