Guanosin-5'-diphosphat Natriumsalz, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Sodium ((2R,3S,4R,5R)-5-(2-amino-6-oxo-1H-purin-9(6H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl diphosphate, Guanosine 5'-diphosphate sodium
M-Clarity™ Qualitätsstufe = MQ200
G7127-100MG
G7127-10MG
G7127-25MG
G7127-500MG
G7127-1G
SIALG7127-100MGEA
200
EUR
SIALG7127-100MG
SIALG7127-10MG
SIALG7127-25MG
SIALG7127-500MG
SIALG7127-1G
Guanosin-5'-diphosphat Natriumsalz, Sigma-Aldrich®
Guanosin-5'-diphosphat Natriumsalz
Formel:
C₁₀H₁₄N₅NaO₁₁P₂ Molecular Weight: 509,1460 g/mol |
MDL:
MFCD00070121 CAS-Nummer: 43139-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...