Guanosin-5'-triphosphat Trinatriumsalz Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
GTP trisodium salt hydrate
51120-100MG
51120-25MG
51120-1G
SIAL51120-100MGEA
147
EUR
SIAL51120-100MG
SIAL51120-25MG
SIAL51120-1G
Guanosin-5'-triphosphat Trinatriumsalz Hydrat, Sigma-Aldrich®
Guanosin-5'-triphosphat Trinatriumsalz Hydrat
Formel:
C₁₀H₁₃N₅Na₃O₁₄P₃·H₂O Molecular Weight: 523,18 g/mol (free acid) Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077781 CAS-Nummer: 36051-31-7 EINECS: 252-847-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...